Product Details

SNP ID
rs11746282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:142780722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGAGTTTTTATCTTTGTGCGGTG[C/T]GTGTAACATTCAGGAGATTCGGCAG
Phenotype
MIM: 605370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP26 PubMed Links

Gene Details

Gene
ARHGAP26
Gene Name
Rho GTPase activating protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135608.1 Intron NP_001129080.1
NM_015071.4 Intron NP_055886.1
XM_005268398.4 Intron XP_005268455.1
XM_005268399.4 Intron XP_005268456.1
XM_005268400.4 Intron XP_005268457.1
XM_005268402.4 Intron XP_005268459.1
XM_006714774.3 Intron XP_006714837.1
XM_011537610.2 Intron XP_011535912.1
XM_011537611.2 Intron XP_011535913.1
XM_011537612.2 Intron XP_011535914.1
XM_017009247.1 Intron XP_016864736.1
XM_017009248.1 Intron XP_016864737.1
XM_017009249.1 Intron XP_016864738.1
XM_017009250.1 Intron XP_016864739.1

View Full Product Details