Product Details
- SNP ID
-
rs986494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:148069504 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAAGAGAGAGAAAGAGAGAGAAAG[C/G]CTTATGCAATTATTGAGAAATTTCC
- Phenotype
-
MIM: 605010
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SPINK5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199650759] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPINK5
- Gene Name
- serine peptidase inhibitor, Kazal type 5
View Full Product Details