Product Details
- SNP ID
-
rs1389832
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:35858619 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAGCTCTATGTAATTTCAAAACCAT[A/G]GTAATTATAACACCGTCAATTGACA
- Phenotype
-
MIM: 146661
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL7R
PubMed Links
Gene Details
- Gene
- IL7R
- Gene Name
- interleukin 7 receptor
View Full Product Details