Product Details

SNP ID
rs11598469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102225872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACTCACAACCGCAGACTCGAAG[A/C]TAGGAGGTGGAAGGGACTACAGAAG
Phenotype
MIM: 611815 MIM: 602669
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELOVL3 PubMed Links

Gene Details

Gene
ELOVL3
Gene Name
ELOVL fatty acid elongase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152310.2 1106 Intron NP_689523.1
XM_011540245.2 1106 UTR 5 XP_011538547.1
Gene
PITX3
Gene Name
paired like homeodomain 3
There are no transcripts associated with this gene.

View Full Product Details