Product Details

SNP ID
rs11974206
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:23715709 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCTCTTGTTTCTCAGGGTCAGTT[A/T]AAATTCAGTTCCTGTTCTATGCTGA
Phenotype
MIM: 605790
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STK31 PubMed Links

Gene Details

Gene
STK31
Gene Name
serine/threonine kinase 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260504.1 Intron NP_001247433.1
NM_001260505.1 Intron NP_001247434.1
NM_031414.4 Intron NP_113602.2
NM_032944.3 Intron NP_116562.2
XM_006715758.3 Intron XP_006715821.1
XM_011515449.2 Intron XP_011513751.2
XM_011515450.1 Intron XP_011513752.1
XM_011515452.2 Intron XP_011513754.2

View Full Product Details