Product Details

SNP ID
rs10954771
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:31001534 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCATTGTCCATGTTGTGAGTACC[C/T]GTACTGTATGCTCTTTGGGGCCAAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PURG PubMed Links

Gene Details

Gene
PURG
Gene Name
purine rich element binding protein G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015508.2 Intron NP_001015508.1
NM_001323311.1 Intron NP_001310240.1
NM_001323312.1 Intron NP_001310241.1
NM_013357.2 Intron NP_037489.1

View Full Product Details