Product Details

SNP ID
rs3740109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45455633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTACAGGAACATTGGCAGAGC[C/T]GGTGGCCACACCAGATGGGAGTACC
Phenotype
MIM: 152390 MIM: 613335
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOX5 PubMed Links

Gene Details

Gene
ALOX5
Gene Name
arachidonate 5-lipoxygenase
There are no transcripts associated with this gene.

Gene
LOC102724323
Gene Name
uncharacterized LOC102724323
There are no transcripts associated with this gene.

Gene
MARCH8
Gene Name
membrane associated ring-CH-type finger 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002266.2 5079 UTR 3 NP_001002266.1
NM_001282866.1 5079 UTR 3 NP_001269795.1
NM_145021.5 5079 UTR 3 NP_659458.2
XM_005271804.2 5079 UTR 3 XP_005271861.1
XM_006717704.2 5079 UTR 3 XP_006717767.1
XM_011539492.2 5079 UTR 3 XP_011537794.1
XM_011539493.2 5079 UTR 3 XP_011537795.1
XM_011539494.2 5079 UTR 3 XP_011537796.1
XM_011539495.1 5079 UTR 3 XP_011537797.1
XM_017015894.1 5079 UTR 3 XP_016871383.1

View Full Product Details