Product Details
- SNP ID
-
rs6707421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:38670639 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCTGTGATATGTGTCTAACCAT[A/G]TCTGACCACATATTTGATACGTCTC
- Phenotype
-
MIM: 137030
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GALM
PubMed Links
Gene Details
- Gene
- GALM
- Gene Name
- galactose mutarotase
View Full Product Details