Product Details
- SNP ID
-
rs1033973
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
57
- Location
-
Chr.1:68101310 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATAAATGTTCATCATATTTGCTAT[C/T]ATTATTATTATTATTATAGGCATCA
- Phenotype
-
MIM: 615406
MIM: 611514
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNG12-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs372414383] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNG12-AS1
- Gene Name
- GNG12 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- WLS
- Gene Name
- wntless Wnt ligand secretion mediator
View Full Product Details