Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001100623.1 | 583 | Intron | NP_001094093.1 | ||
NM_001267062.1 | 583 | Intron | NP_001253991.1 | ||
NM_001267063.1 | 583 | Intron | NP_001253992.1 | ||
NM_001267064.1 | 583 | Intron | NP_001253993.1 | ||
NM_001267065.1 | 583 | Intron | NP_001253994.1 | ||
NM_001267066.1 | 583 | Intron | NP_001253995.1 | ||
NM_001267067.1 | 583 | Intron | NP_001253996.1 | ||
NM_001267068.1 | 583 | Intron | NP_001253997.1 | ||
NM_001309448.1 | 583 | Intron | NP_001296377.1 | ||
NM_001309450.1 | 583 | Intron | NP_001296379.1 | ||
NM_001309451.1 | 583 | Intron | NP_001296380.1 | ||
NM_001309452.1 | 583 | Intron | NP_001296381.1 | ||
NM_017958.2 | 583 | Intron | NP_060428.2 | ||
XM_011511385.2 | 583 | Intron | XP_011509687.1 | ||
XM_011511386.2 | 583 | Intron | XP_011509688.1 | ||
XM_017004408.1 | 583 | Intron | XP_016859897.1 | ||
XM_017004409.1 | 583 | Intron | XP_016859898.1 | ||
XM_017004410.1 | 583 | Intron | XP_016859899.1 | ||
XM_017004411.1 | 583 | UTR 5 | XP_016859900.1 | ||
XM_017004412.1 | 583 | UTR 5 | XP_016859901.1 | ||
XM_017004413.1 | 583 | Intron | XP_016859902.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001277406.1 | 583 | Intron | NP_001264335.1 | ||
XM_011511670.2 | 583 | Intron | XP_011509972.1 | ||
XM_017004732.1 | 583 | Intron | XP_016860221.1 | ||
XM_017004733.1 | 583 | Intron | XP_016860222.1 | ||
XM_017004734.1 | 583 | Intron | XP_016860223.1 | ||
XM_017004735.1 | 583 | Intron | XP_016860224.1 |