Product Details

SNP ID
rs11853719
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24987946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTTAAATTATCTACGTGATAT[A/G]AGGAAACGAGCTTATTTTTGATAAT
Phenotype
MIM: 600162 MIM: 182279
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PWAR5 PubMed Links
Additional Information
For this assay, SNP(s) [rs78165937] are located under a probe and SNP(s) [rs147947212] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PWAR5
Gene Name
Prader Willi/Angelman region RNA 5
There are no transcripts associated with this gene.

Gene
PWARSN
Gene Name
Prader Willi/Angelman region RNA, SNRPN neighbor
There are no transcripts associated with this gene.

Gene
SNORD107
Gene Name
small nucleolar RNA, C/D box 107
There are no transcripts associated with this gene.

Gene
SNORD108
Gene Name
small nucleolar RNA, C/D box 108
There are no transcripts associated with this gene.

Gene
SNORD64
Gene Name
small nucleolar RNA, C/D box 64
There are no transcripts associated with this gene.

Gene
SNRPN
Gene Name
small nuclear ribonucleoprotein polypeptide N
There are no transcripts associated with this gene.

Gene
SNURF
Gene Name
SNRPN upstream reading frame
There are no transcripts associated with this gene.

View Full Product Details