Product Details

SNP ID
rs6785639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122800236 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGACTGAGCCTTGGGGCATTCCA[A/T]CATTCAGAGGTTGGGGAGATAAGAA
Phenotype
MIM: 602773 MIM: 608263
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DIRC2 PubMed Links

Gene Details

Gene
DIRC2
Gene Name
disrupted in renal carcinoma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032839.2 Intron NP_116228.1
Gene
HSPBAP1
Gene Name
HSPB1 associated protein 1
There are no transcripts associated with this gene.

View Full Product Details