Product Details

SNP ID
rs1106863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126988533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTTCCCCAGGGCTGAAGCTCC[C/T]GGCACCATGATGCTCACCCCAGCAG
Phenotype
MIM: 601055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNA1 PubMed Links

Gene Details

Gene
PLXNA1
Gene Name
plexin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032242.3 635 Intron NP_115618.3
XM_011512908.2 635 UTR 5 XP_011511210.1
XM_011512909.1 635 UTR 5 XP_011511211.1

View Full Product Details