Product Details

SNP ID
rs4147384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159271060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCATCATTTAACCATGTTATTC[A/C]CTGTTGATTATAAATAATTTTTTAT
Phenotype
MIM: 609867
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBLCP1 PubMed Links

Gene Details

Gene
UBLCP1
Gene Name
ubiquitin like domain containing CTD phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145049.3 Intron NP_659486.2

View Full Product Details