Product Details
- SNP ID
-
rs11773176
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:2564844 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTGATCAAGTTTTTCTAACTCCT[C/T]GTTGATCTTCTGCCTCGCTCTCCTG
- Phenotype
-
MIM: 614506
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BRAT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116093568] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BRAT1
- Gene Name
- BRCA1 associated ATM activator 1
There are no transcripts associated with this gene.
- Gene
- IQCE
- Gene Name
- IQ motif containing E
View Full Product Details