Product Details

SNP ID
rs12700069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2568261 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAAAAACACAATATAAAATTACC[A/T]TTGCACTATGGGTATAATGTATATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IQCE PubMed Links

Gene Details

Gene
IQCE
Gene Name
IQ motif containing E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287499.1 Intron NP_001274428.1
NM_001287500.1 Intron NP_001274429.1
NM_001287501.1 Intron NP_001274430.1
NM_001287502.1 Intron NP_001274431.1
NM_152558.4 Intron NP_689771.3
XM_006715676.2 Intron XP_006715739.1
XM_011515242.1 Intron XP_011513544.1
XM_011515243.1 Intron XP_011513545.1
XM_011515244.2 Intron XP_011513546.1
XM_017011902.1 Intron XP_016867391.1
XM_017011903.1 Intron XP_016867392.1

View Full Product Details