Product Details
- SNP ID
-
rs2576974
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:103844104 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGAGCCATTCCATATTTCAGCATT[A/G]TTTTGTGTCTTCCGTTGTATCTTTT
- Phenotype
-
MIM: 611076
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NT5DC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77247938] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NT5DC3
- Gene Name
- 5'-nucleotidase domain containing 3
There are no transcripts associated with this gene.
- Gene
- TTC41P
- Gene Name
- tetratricopeptide repeat domain 41, pseudogene
There are no transcripts associated with this gene.
View Full Product Details