Product Details

SNP ID
rs12427129
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:53973906 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCCACTCCGTGGCCAAGGAGCCGG[C/T]CAAAGGAGCCGCCCCCAGTAGGTAG
Phenotype
MIM: 611400 MIM: 605559
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HOTAIR PubMed Links

Gene Details

Gene
HOTAIR
Gene Name
HOX transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXC11
Gene Name
homeobox C11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014212.3 781 Missense Mutation GCC,GTC A222V NP_055027.1

View Full Product Details