Product Details
- SNP ID
-
rs35757523
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12107227 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATCTTCAATTATGGATCCACCCTGA[C/T]ATATTTGAAAGAGCTGCAGTATTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF788
PubMed Links
Gene Details
- Gene
- ZNF788
- Gene Name
- zinc finger family member 788
There are no transcripts associated with this gene.
View Full Product Details