Product Details

SNP ID
rs841198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4618565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCTCTGAATATGATTCCCGCT[A/G]TAATAATTACCACTAGGACGACAAC
Phenotype
MIM: 607559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGRN1 PubMed Links

Gene Details

Gene
MGRN1
Gene Name
mahogunin ring finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142289.2 Intron NP_001135761.2
NM_001142290.2 Intron NP_001135762.1
NM_001142291.2 Intron NP_001135763.2
NM_015246.3 Intron NP_056061.1
XM_005255217.1 Intron XP_005255274.1
XM_005255218.1 Intron XP_005255275.1
XM_005255219.3 Intron XP_005255276.1
XM_005255220.3 Intron XP_005255277.1
XM_005255221.3 Intron XP_005255278.1
XM_011522441.1 Intron XP_011520743.1
XM_011522442.2 Intron XP_011520744.1
XM_017023091.1 Intron XP_016878580.1
XM_017023092.1 Intron XP_016878581.1
XM_017023093.1 Intron XP_016878582.1
Gene
UBALD1
Gene Name
UBA like domain containing 1
There are no transcripts associated with this gene.

View Full Product Details