Product Details
- SNP ID
-
rs8059151
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:4621728 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACATGTACATGTACGTGAGTTACG[G/T]GATTCAATGGAAAGAAATATAAAAT
- Phenotype
-
MIM: 607559
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MGRN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74434023] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MGRN1
- Gene Name
- mahogunin ring finger 1
- Gene
- UBALD1
- Gene Name
- UBA like domain containing 1
There are no transcripts associated with this gene.
View Full Product Details