Product Details

SNP ID
rs10810812
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:17586103 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAACAAAATATTTTCATGCTATACT[A/G]AATCATTAGCTGATGACAAATGCTT
Phenotype
MIM: 604465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH3GL2 PubMed Links

Gene Details

Gene
SH3GL2
Gene Name
SH3 domain containing GRB2 like 2, endophilin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003026.3 Intron NP_003017.1
XM_011518005.2 Intron XP_011516307.1

View Full Product Details