Product Details
- SNP ID
-
rs3746444
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:34990448 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTTAACTCCTCTCCACGTGAAC[G/A]TCACAGCAAGTCTGTGCTGCTTCCC
- Phenotype
-
MIM: 613614
MIM: 609928
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR499A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7267163] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR499A
- Gene Name
- microRNA 499a
There are no transcripts associated with this gene.
- Gene
- MIR499B
- Gene Name
- microRNA 499b
There are no transcripts associated with this gene.
- Gene
- MYH7B
- Gene Name
- myosin heavy chain 7B
View Full Product Details