Product Details

SNP ID
rs11617003
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:24685665 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAGGCAAATGTGTATGGAAGTGTCC[C/T]GAGATTCACTCTATCCCCCTGGGAT
Phenotype
MIM: 182360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP12A PubMed Links

Gene Details

Gene
ATP12A
Gene Name
ATPase H+/K+ transporting non-gastric alpha2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185085.1 Intron NP_001172014.1
NM_001676.5 Intron NP_001667.4

View Full Product Details