Product Details
- SNP ID
-
rs2896489
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103642406 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAATAACCTTGATAGTTTTGAGAC[A/G]TATGGGCAAGGTATTTTGTACAATG
- Phenotype
-
MIM: 600025
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLC1
PubMed Links
Gene Details
- Gene
- KLC1
- Gene Name
- kinesin light chain 1
View Full Product Details