Product Details
- SNP ID
-
rs108621
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:75013934 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGCATGATCGCGACTGGCCAGCA[C/T]ACTGGCCGGTTCTCTCTGTTAGCAG
- Phenotype
-
MIM: 606454
MIM: 604395
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF2B2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77514025] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF2B2
- Gene Name
- eukaryotic translation initiation factor 2B subunit beta
There are no transcripts associated with this gene.
- Gene
- MLH3
- Gene Name
- mutL homolog 3
View Full Product Details