Product Details

SNP ID
rs6517574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39874735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTTCTAGTATTTTAAAAAAAAA[C/G]CTAGTTAAAAGGTTACACCTGCTTT
Phenotype
MIM: 601629
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCP4 PubMed Links

Gene Details

Gene
PCP4
Gene Name
Purkinje cell protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006198.2 Intron NP_006189.2

View Full Product Details