Product Details

SNP ID
rs35923562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29211352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAACTTTAGCAACACGACGAA[C/T]TCCAAAGATGCTTTTCTTTTGCTAT
Phenotype
MIM: 191327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UQCRFS1 PubMed Links

Gene Details

Gene
UQCRFS1
Gene Name
ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006003.2 Intron NP_005994.2

View Full Product Details