Product Details

SNP ID
rs17733783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127998669 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGCGAAGTACTTCAGAAAGCCT[A/G]GAGAGAAGAGAGGGCCCTCAAATCC
Phenotype
MIM: 602181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SND1 PubMed Links

Gene Details

Gene
SND1
Gene Name
staphylococcal nuclease and tudor domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011987.1 Intron XP_016867476.1
Gene
SND1-IT1
Gene Name
SND1 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details