Product Details

SNP ID
rs16865904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224379399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGACTGTGACGGCAAATAAACAA[C/T]CATTCCCAGCACCTTTAGACTTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM124B PubMed Links

Gene Details

Gene
FAM124B
Gene Name
family with sequence similarity 124 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122779.1 1893 UTR 3 NP_001116251.1
NM_024785.2 1893 UTR 3 NP_079061.2

View Full Product Details