Product Details

SNP ID
rs440081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142835655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATTTGGGGGTCTATTTCTTATT[C/T]GGAGGTGTGTTCTTAAGTTCCAAAC
Phenotype
MIM: 602370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GML PubMed Links

Gene Details

Gene
GML
Gene Name
glycosylphosphatidylinositol anchored molecule like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002066.2 Intron NP_002057.1

View Full Product Details