Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001289174.1 | Intron | NP_001276103.1 | |||
NM_001289175.1 | Intron | NP_001276104.1 | |||
NM_001289176.1 | Intron | NP_001276105.1 | |||
NM_022553.5 | Intron | NP_072047.4 | |||
XM_011514797.1 | Intron | XP_011513099.1 | |||
XM_011514798.2 | Intron | XP_011513100.1 | |||
XM_011514799.1 | Intron | XP_011513101.1 | |||
XM_017011177.1 | Intron | XP_016866666.1 | |||
XM_017011178.1 | Intron | XP_016866667.1 | |||
XM_017011179.1 | Intron | XP_016866668.1 |