Product Details

SNP ID
rs767623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26526494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGACCTCAAATATACACTTTT[C/T]TTATGGACACAGAGCAAATGACAGT
Phenotype
MIM: 603126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPST2 PubMed Links

Gene Details

Gene
TPST2
Gene Name
tyrosylprotein sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008566.1 Intron NP_001008566.1
NM_003595.3 Intron NP_003586.3

View Full Product Details