Product Details

SNP ID
rs763780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52236941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGATATGCACCTCTTACTGCACA[C/T]GGTGGATGACAGGGGTGACGCAGGT
Phenotype
MIM: 606496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17F PubMed Links

Gene Details

Gene
IL17F
Gene Name
interleukin 17F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052872.3 619 Missense Mutation CAT,CGT H161R NP_443104.1
XM_011514276.1 619 Missense Mutation CAT,CGT H161R XP_011512578.1

View Full Product Details