Product Details

SNP ID
rs1076726
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:15152787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGCGGAGGCAGAGCCGCGGGT[A/G]AGAGGCTTCCGCGCCGACTTCGGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM51 PubMed Links

Gene Details

Gene
TMEM51
Gene Name
transmembrane protein 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136216.1 370 Intron NP_001129688.1
NM_001136217.1 370 Intron NP_001129689.1
NM_001136218.1 370 Intron NP_001129690.1
NM_001319665.1 370 Intron NP_001306594.1
NM_018022.2 370 Intron NP_060492.1
XM_005245919.1 370 Intron XP_005245976.1
XM_011541676.1 370 Intron XP_011539978.1
XM_017001590.1 370 UTR 5 XP_016857079.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details