Product Details

SNP ID
rs139105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44173466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCATAGCAGGTTTCAGGTGCGTC[A/G]AATTTTACGAAATGATGATAAAATT
Phenotype
MIM: 608121 MIM: 608122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARVB PubMed Links

Gene Details

Gene
PARVB
Gene Name
parvin beta
There are no transcripts associated with this gene.

Gene
PARVG
Gene Name
parvin gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137605.2 Intron NP_001131077.1
NM_022141.6 Intron NP_071424.1
XM_005261702.3 Intron XP_005261759.1
XM_011530302.2 Intron XP_011528604.1
XM_017028907.1 Intron XP_016884396.1
XM_017028908.1 Intron XP_016884397.1

View Full Product Details