Product Details

SNP ID
rs621627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42695549 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATAGTCATCACCTGTTTCTTCAC[C/T]GTGGAGTGAGCATTTTAAAAATGAA
Phenotype
MIM: 179605 MIM: 609134
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPH2 PubMed Links

Gene Details

Gene
PRPH2
Gene Name
peripherin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000322.4 Intron NP_000313.2
Gene
UBR2
Gene Name
ubiquitin protein ligase E3 component n-recognin 2
There are no transcripts associated with this gene.

View Full Product Details