Product Details
- SNP ID
-
rs1712
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:152970730 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAAAGCTTTAGAAAGGTGACAA[C/T]ACATTTAATAGACTCCAGAAGTTGA
- Phenotype
-
MIM: 606013
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO5
PubMed Links
Gene Details
- Gene
- FBXO5
- Gene Name
- F-box protein 5
View Full Product Details