Product Details

SNP ID
rs739989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50353836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCGGGTCCCACCTGCCCCACC[C/T]CCGGAAGCCCCAGGCCGCCAGAACC
Phenotype
MIM: 608125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XYLT2 PubMed Links

Gene Details

Gene
XYLT2
Gene Name
xylosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022167.3 765 Silent Mutation CCC,CCT P114P NP_071450.2
XM_005257572.4 765 Silent Mutation CCC,CCT P82P XP_005257629.1

View Full Product Details