Product Details

SNP ID
rs266811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1397048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCTGTGTCCATGGGATGGGC[C/G]GGAGGTGAGGGAGCAGCCGCTGGAA
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000156.5 1116 UTR 3 NP_000147.1
NM_138924.2 1116 Intron NP_620279.1
Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
There are no transcripts associated with this gene.

View Full Product Details