Product Details

SNP ID
rs1076295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30365248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGGACAGGGGAGCATGATTCAGA[C/T]AGAGGGGACAGATGGGCGAGGATTT
Phenotype
MIM: 605595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC157 PubMed Links

Gene Details

Gene
CCDC157
Gene Name
coiled-coil domain containing 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017437.3 Intron NP_001017437.2
NM_001318334.1 Intron NP_001305263.1
NM_001318335.1 Intron NP_001305264.1
XM_006724278.3 Intron XP_006724341.1
XM_006724279.3 Intron XP_006724342.1
XM_006724280.3 Intron XP_006724343.1
XM_011530263.2 Intron XP_011528565.1
XM_011530266.2 Intron XP_011528568.1
XM_011530267.2 Intron XP_011528569.1
XM_011530268.2 Intron XP_011528570.1
XM_011530269.2 Intron XP_011528571.1
XM_011530270.2 Intron XP_011528572.1
Gene
KIAA1656
Gene Name
KIAA1656 protein
There are no transcripts associated with this gene.

Gene
SF3A1
Gene Name
splicing factor 3a subunit 1
There are no transcripts associated with this gene.

View Full Product Details