Product Details

SNP ID
rs72547557
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.17:19651626 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TTTATGCTTGAGAATCTTCCTGAAT[A/G]GGTTACTGCTAAACCAGTTAAGAAG
Phenotype
MIM: 609523
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH3A2 PubMed Links

Gene Details

Gene
ALDH3A2
Gene Name
aldehyde dehydrogenase 3 family member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000382.2 362 Nonsense Mutation TAG,TGG *78W NP_000373.1
NM_001031806.1 362 Nonsense Mutation TAG,TGG *78W NP_001026976.1
XM_011523732.1 362 Nonsense Mutation TAG,TGG *78W XP_011522034.1
XM_011523733.1 362 Nonsense Mutation TAG,TGG *78W XP_011522035.1
XM_017024355.1 362 Nonsense Mutation TAG,TGG *78W XP_016879844.1
XM_017024356.1 362 Nonsense Mutation TAG,TGG *78W XP_016879845.1
XM_017024357.1 362 Nonsense Mutation TAG,TGG *78W XP_016879846.1
XM_017024358.1 362 Nonsense Mutation TAG,TGG *78W XP_016879847.1

View Full Product Details