Product Details

SNP ID
rs763473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76681647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCCCTGCAGACCTCAAATGGA[C/G]GGGTGGGTCCTGATCTATCAGAGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105274304 PubMed Links

Gene Details

Gene
LOC105274304
Gene Name
uncharacterized LOC105274304
There are no transcripts associated with this gene.

Gene
MXRA7
Gene Name
matrix remodeling associated 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008528.2 Intron NP_001008528.1
NM_001008529.2 Intron NP_001008529.1
NM_198530.3 Intron NP_940932.2
XM_005257382.2 Intron XP_005257439.1
XM_005257383.2 Intron XP_005257440.1
XM_005257384.2 Intron XP_005257441.1
XM_011524830.1 Intron XP_011523132.1

View Full Product Details