Product Details

SNP ID
rs42036
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:92612137 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTTTATATGTAGGTTGTTTTGACT[C/G]CACCCTGTGCAACAGTTCCCAAAAT
Phenotype
MIM: 603368
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK6 PubMed Links

Gene Details

Gene
CDK6
Gene Name
cyclin dependent kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145306.1 4621 UTR 3 NP_001138778.1
NM_001259.6 4621 UTR 3 NP_001250.1
XM_006715835.2 4621 UTR 3 XP_006715898.1

View Full Product Details