Product Details
- SNP ID
-
rs757719
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:101171466 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTTAAGCGTCGTAGAGATCCAG[A/C]GAGGATGAGGATGAGGGGCTAGAGA
- Phenotype
-
MIM: 615783
MIM: 602186
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NAT16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75082775] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NAT16
- Gene Name
- N-acetyltransferase 16 (putative)
- Gene
- VGF
- Gene Name
- VGF nerve growth factor inducible
There are no transcripts associated with this gene.
View Full Product Details