Product Details

SNP ID
rs740104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101173066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGATCCTAGGGGAGGCAGGGTC[C/G]CCGAAGTTGGGAAGTAAGGACTAGG
Phenotype
MIM: 615783 MIM: 602186
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAT16 PubMed Links

Gene Details

Gene
NAT16
Gene Name
N-acetyltransferase 16 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198571.2 Intron NP_940973.2
XM_005250325.1 Intron XP_005250382.1
XM_011516168.1 Intron XP_011514470.1
Gene
VGF
Gene Name
VGF nerve growth factor inducible
There are no transcripts associated with this gene.

View Full Product Details