Product Details

SNP ID
rs243538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:148707287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTTGTAGCACAGTGTGGGAAGT[A/G]CCATAGAGGCCAGGATCCTAATGAG
Phenotype
MIM: 603134
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL1 PubMed Links

Gene Details

Gene
CUL1
Gene Name
cullin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003592.2 Intron NP_003583.2
XM_005250060.4 Intron XP_005250117.1
XM_011516629.2 Intron XP_011514931.1
XM_011516630.2 Intron XP_011514932.1
XM_011516631.2 Intron XP_011514933.1
XM_011516632.2 Intron XP_011514934.1
XM_017012723.1 Intron XP_016868212.1

View Full Product Details