Product Details

SNP ID
rs714775
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:6510236 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACATTTTGTCAGCCACATGATGATA[C/T]CAAGGCTGTTGTGATTCAGTTGGTT
Phenotype
MIM: 615638 MIM: 615639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCAPD2 PubMed Links

Gene Details

Gene
NCAPD2
Gene Name
non-SMC condensin I complex subunit D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014865.3 Intron NP_055680.3
Gene
SCARNA10
Gene Name
small Cajal body-specific RNA 10
There are no transcripts associated with this gene.

View Full Product Details