Product Details

SNP ID
rs304505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89378825 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGCTGGCTTAATAAAAGCAGCA[A/G]TGCTCTTAAGGAAATAAACGAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFIT1B PubMed Links

Gene Details

Gene
IFIT1B
Gene Name
interferon induced protein with tetratricopeptide repeats 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010987.2 Intron NP_001010987.1

View Full Product Details