Product Details

SNP ID
rs1167
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:87966831 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAATGAGTGCTCAGTGATGTGAAG[C/T]ACACAGGAGTCCCTCAGGGCAAAAG
Phenotype
MIM: 610076
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK20 PubMed Links

Gene Details

Gene
CDK20
Gene Name
cyclin dependent kinase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039803.2 1982 UTR 3 NP_001034892.1
NM_001170639.1 1982 UTR 3 NP_001164110.1
NM_001170640.1 1982 UTR 3 NP_001164111.1
NM_012119.4 1982 UTR 3 NP_036251.2
NM_178432.3 1982 UTR 3 NP_848519.1
XM_017014561.1 1982 UTR 3 XP_016870050.1
XM_017014562.1 1982 UTR 3 XP_016870051.1
XM_017014563.1 1982 UTR 3 XP_016870052.1
XM_017014564.1 1982 UTR 3 XP_016870053.1

View Full Product Details